missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYN Mouse anti-Human, Mouse, Clone: 3C7F2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60211-1-IG-150UL
This item is not returnable.
View return policy
Description
Prohibitin is an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3′ untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3′ untranslated region may function as a trans-acting regulatory RNA.Specifications
| LYN | |
| Monoclonal | |
| 1.08 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07948, P25911 | |
| Lyn | |
| LYN Fusion Protein Ag12723 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3C7F2 | |
| Unconjugated | |
| LYN | |
| JTK8, LYN, Tyrosine protein kinase Lyn | |
| Mouse | |
| Protein A | |
| RUO | |
| 17096, 4067 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction