missing translation for 'onlineSavingsMsg'
Learn More

LYN Mouse anti-Human, Mouse, Clone: 3C7F2, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen LYN
Clone 3C7F2
Concentration 1.08 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16807225
View Documents
Proteintech
60211-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16897215
View Documents
Proteintech
60211-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Prohibitin is an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3′ untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3′ untranslated region may function as a trans-acting regulatory RNA.
Specifications

Specifications

LYN
1.08 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
JTK8, LYN, Tyrosine protein kinase Lyn
Lyn
IgG2a
Protein A
Antibody
3C7F2
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse
P07948, P25911
17096, 4067
LYN Fusion Protein Ag12723
Primary
-20°C
LYN
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.