missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lyl1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | Lyl1 |
|---|---|
| Dilution | Western Blot 3-5ug/ml, Simple Western 1:10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18123043
|
Novus Biologicals
NBP2-27188SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18111312
|
Novus Biologicals
NBP2-27188 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lyl1 Polyclonal specifically detects Lyl1 in Human, Porcine, Canine, Primate samples. It is validated for Western Blot, Simple Western.Specifications
| Lyl1 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| No buffer with No Preservative | |
| 4066 | |
| 235 RSQPAPPADPDGSPGG 250 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 3-5ug/ml, Simple Western 1:10 | |
| Polyclonal | |
| Rabbit | |
| Human, Pig, Canine, Primate | |
| BHLHA18, bHLHa18Class A basic helix-loop-helix protein 18, lymphoblastic leukemia derived sequence 1, Lymphoblastic leukemia-derived sequence 1, protein lyl-1 | |
| LYL1 | |
| IgG | |
| Affinity Purified | |
| Dog, Human, Orangutan, Wild pig. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title