missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lyl1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27188SS
This item is not returnable.
View return policy
Description
Lyl1 Polyclonal specifically detects Lyl1 in Human, Porcine, Canine, Primate samples. It is validated for Western Blot, Simple Western.
Specifications
| Lyl1 | |
| Polyclonal | |
| Western Blot 3-5ug/ml, Simple Western 1:10 | |
| BHLHA18, bHLHa18Class A basic helix-loop-helix protein 18, lymphoblastic leukemia derived sequence 1, Lymphoblastic leukemia-derived sequence 1, protein lyl-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4066 | |
| Human, Pig, Canine, Primate | |
| IgG |
| Western Blot | |
| Unconjugated | |
| No buffer with No Preservative | |
| LYL1 | |
| 235 RSQPAPPADPDGSPGG 250 | |
| 0.025 mg | |
| Primary | |
| Dog, Human, Orangutan, Wild pig. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction