missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat, anti-Mouse, APC, Clone: 1A8, STEMCELL Technologies
Rat Monoclonal Antibody
£162.00 - £583.20
Specifications
| Antigen | Ly-6G |
|---|---|
| Clone | 1A8 |
| Applications | Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Functional Assay |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17178631
|
Stemcell Technologies
60031AZ.1 |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17168631
|
Stemcell Technologies
60031AZ |
100 μg |
£583.20
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The 1A8 antibody reacts with Ly-6G, a 21 - 25 kDa GPI-anchored protein, which together with the structurally related Ly-6C protein comprises the granulocyte receptor-1 antigen (Gr-1)
- Gr-1 is expressed on monocytes, neutrophils and subsets of macrophages, plasmacytoid dendritic cells, and T cells
- Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
- In the periphery, Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
- The 1A8 antibody binds specifically to Ly-6G, whereas another commonly used antibody, clone RB6-8C5, binds to both Ly-6G and Ly-6C
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Specifications
| Ly-6G | |
| Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Functional Assay | |
| APC | |
| Mouse | |
| Primary |
| 1A8 | |
| Monoclonal | |
| Rat | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title