Learn More
Ly-6G Rat, anti-Mouse, APC, Clone: 1A8, STEMCELL Technologies
Rat Monoclonal Antibody
Brand: Stemcell Technologies 60031AZ.1
This item is not returnable.
View return policy
Description
- The 1A8 antibody reacts with Ly-6G, a 21 - 25 kDa GPI-anchored protein, which together with the structurally related Ly-6C protein comprises the granulocyte receptor-1 antigen (Gr-1)
- Gr-1 is expressed on monocytes, neutrophils and subsets of macrophages, plasmacytoid dendritic cells, and T cells
- Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
- In the periphery, Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
- The 1A8 antibody binds specifically to Ly-6G, whereas another commonly used antibody, clone RB6-8C5, binds to both Ly-6G and Ly-6C
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Specifications
| Ly-6G | |
| Monoclonal | |
| APC | |
| 25 μg | |
| Mouse | |
| Granulocytes and Subsets, Myeloid Cells |
| Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Functional Assay | |
| 1A8 | |
| Rat | |
| Primary | |
| IgG2a κ |