missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LY 456236 hydrochloride
GREENER_CHOICE

Product Code. 18065662
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18065662

Brand: Tocris Bioscience™ 2390/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective mGlu1 antagonist

Selective mGlu1 receptor antagonist (IC50 values are 143 nM and > 10 μM for mGlu1 and mGlu5 receptors respectively). Reduces hyperalgesic behavior induced by formalin in both mouse and rat with ED50 values of 28 and 16.3 mg/kg respectively.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338736-46-2
Molecular Formula C16H16ClN3O2

Specifications

Quantity 50 mg
Formula Weight 317.77
Purity >99%
Molecular Weight (g/mol) 317.773
Chemical Name or Material LY 456236 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.