accessibility menu, dialog, popup

UserName

CAS RN 338736-46-2

H3CONHNNOCH3HCl

CAS RN 338736-46-2

IUPAC Name: 6-methoxy-N-(4-methoxyphenyl)quinazolin-4-amine hydrochloride
Synonyms: 6-methoxy-N-(4-methoxyphenyl)quinazolin-4-amine hydrochloride
Molecular Weight (g/mol): 317.77
Molecular Formula: C16H16ClN3O2
InChi Key: AVKFOWUSTVWZQN-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(NC2=C3C=C(OC)C=CC3=NC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results