missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LY 456236 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective mGlu1 receptor antagonist (IC50 values are 143 nM and > 10 μM for mGlu1 and mGlu5 receptors respectively). Reduces hyperalgesic behavior induced by formalin in both mouse and rat with ED50 values of 28 and 16.3 mg/kg respectively.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 317.77 |
| Purity | >99% |
| Molecular Weight (g/mol) | 317.773 |
| Chemical Name or Material | LY 456236 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction