missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR alpha/NR1H3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£185.00 - £417.00
Specifications
| Antigen | LXR |
|---|---|
| Concentration | 1.04 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Simple Western |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18044894
|
Novus Biologicals
NB400-157SS |
0.025 mL |
£185.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18738513
|
Novus Biologicals
NB400-157 |
0.1 mL |
£417.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LXR alpha/NR1H3 Polyclonal specifically detects LXR alpha/NR1H3 in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Simple Western.Specifications
| LXR | |
| Western Blot 0.5-2 ug/ml, Simple Western | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
| NR1H3 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, porcine and monkey. Immunogen sequence has 92% identity to rat and 90% identity to chicken. |
| 1.04 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Pig, Primate | |
| Q13133 | |
| 10062 | |
| A synthetic peptide made to an internal portion of the human LXR protein sequence (between residues 50-150). [UniProt# Q13133] | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title