missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR alpha/NR1H3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-157
This item is not returnable.
View return policy
Description
LXR alpha/NR1H3 Polyclonal specifically detects LXR alpha/NR1H3 in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Simple Western.
Specifications
| LXR | |
| Polyclonal | |
| Unconjugated | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, porcine and monkey. Immunogen sequence has 92% identity to rat and 90% identity to chicken. | |
| Store at 4C. Do not freeze. |
| Western Blot | |
| 1.04 mg/mL | |
| Western Blot 0.5-2 ug/ml, Simple Western | |
| Q13133 | |
| NR1H3 | |
| A synthetic peptide made to an internal portion of the human LXR protein sequence (between residues 50-150). [UniProt# Q13133] | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators | |
| 10062 | |
| Human, Mouse, Pig, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction