missing translation for 'onlineSavingsMsg'
Learn More

Welch Ilmvac™ LVS301Z Netvac1 Chemical Duty Vacuum Pump

Product Code. 15422055 Shop All Welch Ilmvac Products
This item is not returnable. View return policy

Product Code. 15422055

Brand: Welch Ilmvac™ 11504702

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Laboratory vacuum system
netvac 1, LVS with clamping ring screw fitting A 10.
Conforms to 2006/95/EC Low Voltage Directive; 2006/42/EC Machinery Directive; 2004/108/EC Electromagnetic Compatibility Directive; 94/9/EC ATEX Guideline for use in potentially explosive atmospheres, Appendix III.
• Chemically resistant diaphragm pump MPC 301 Z with gas ballast valve
• Suction-side separator with 500mL round bottom flask
• Pressure-side, insulated intensive cooler with round bottom flask 500mL with safety valve
• One manual regulated connection
• Suction connection DN8 for hose inside diameter 8mm
• Suction-side: clamping ring screw fitting A10
• Assembled and wired ready for connection
TRUSTED_SUSTAINABILITY
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.