missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luteolin, Tocris Bioscience™
Anti-inflammatory, antioxidant and free radical scavenger
£102.92 - £316.51
Chemical Identifiers
CAS | 491-70-3 |
---|---|
Molecular Formula | C15H10O6 |
Molecular Weight (g/mol) | 286.239 |
InChI Key | IQPNAANSBPBGFQ-UHFFFAOYSA-N |
Synonym | luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide |
PubChem CID | 5280445 |
ChEBI | CHEBI:15864 |
IUPAC Name | 2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one |
SMILES | C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18064562
|
Tocris Bioscience
2874/10 |
10 mg |
£102.92
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18064062
|
Tocris Bioscience
2874/50 |
50 mg |
£316.51
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Anti-inflammatory, antioxidant and free radical scavenger. Inhibits LPS-induced TNF-α, IL-6 and inducible nitric oxide production and blocks NF-κB and AP-1 activation. Also inhibits TNF-α -induced COX-2 expression. Antiproliferative and chemopreventative; inhibits proliferation of Lewis lung carcinoma cells in vivo. Inhibits influenza virus replication in vitro by preventing viral entry. Molecular docking studies also indicate binding to SARS-CoV-2 main protease (Mpro or 3CLpro).Chemical Identifiers
491-70-3 | |
286.239 | |
luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide | |
CHEBI:15864 | |
C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O |
C15H10O6 | |
IQPNAANSBPBGFQ-UHFFFAOYSA-N | |
5280445 | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one |
Specifications
491-70-3 | |
luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide | |
C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O | |
286.239 | |
CHEBI:15864 | |
>98% |
C15H10O6 | |
IQPNAANSBPBGFQ-UHFFFAOYSA-N | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one | |
5280445 | |
286.24 | |
Luteolin |
Safety and Handling
Recommended Storage : Store at +4°C