missing translation for 'onlineSavingsMsg'
Learn More

Luteolin- 7-O-Glucuronide, TRC

Product Code. 30133234
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133234 25 mg 25mg
30137541 5 mg 5mg
17981475 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133234 Supplier Toronto Research Chemicals Supplier No. TRCL47551025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29741-10-4
Molecular Formula C21H18O12
Molecular Weight (g/mol) 462.36
Synonym β-D-Glucopyranosiduronic acid, 2-(3,4-dihydroxyphenyl)-5-hydroxy-4-oxo-4H-1-benzopyran-7-yl, Glucopyranosiduronic acid, 2-(3,4-dihydroxyphenyl)-5-hydroxy-4-oxo-4H-1-benzopyran-7-yl, β-D- (8CI), Flavone, 3',4',5,7-tetrahydroxy-, 7-β-D-glucopyranuronoside (8CI), 2-(3,4-Dihydroxyphenyl)-5-hydroxy-4-oxo-4H-1-benzopyran-7-yl β-D-glucopyranosiduronic acid, Cyanidenon-7-O-β-D-glucuronic acid, Luteolin 7-O-glucuronide, Luteolin 7-O-β-D-glucuronide, Luteolin 7-O-β-D-glucuronopyranoside, Luteolin 7-O-β-glucuronide, Luteolin 7-O-β-glucuronopyranoside, Luteolin 7-glucuronide, Luteolin-7-β-D-glucuronide, Cyanidenon-7-O-beta-D-glucuronic Acid, Luteolin 7-O-Glucuronide, Luteolin 7-O-beta-D-Glucuronide, Luteolin 7-O-beta-D-Glucuronopyranoside, Luteolin 7-O-beta-Glucuronide, Luteolin 7-O-beta-Glucuronopyranoside, Luteolin 7-Glucuronide;Luteolin-7-beta-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-[2-(3,4-dihydroxyphenyl)-5-hydroxy-4-oxochromen-7-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES O[C@@H]1[C@@H](O)[C@H](Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)c(O)c4)O[C@@H]([C@H]1O)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H18O12/c22-9-2-1-7(3-10(9)23)13-6-12(25)15-11(24)4-8(5-14(15)32-13)31-21-18(28)16(26)17(27)19(33-21)20(29)30/h1-6,16-19,21-24,26-28H,(H,29,30)/t16-,17-,18+,19-,21+/m0/s1
Formula Weight 462.0798
Chemical Name or Material Luteolin- 7-O-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.