missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Luminoskan™ Microplate Luminometer

Product Code. 15883537 Shop All Thermo Scientific Products
Click to view available options
:
420 mm (16.5 in.) (W) x 420 mm (16.5 in.) (D) x 340 mm (13.4 in.) (H)
This item is not returnable. View return policy

Product Code. 15883537

Brand: Thermo Scientific™ 5300330

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Useful for a variety of luminometric research applications, the Thermo Scientific Luminoskan Microplate Luminometer is a compact and robust instrument offering plate format versatility, fast reading speeds, and up to three dispensers.

This instrument has passed validation at Promega Corporation to be certified as SkanIt software is quick and easy to learn. It offers easy data analysis (with a comprehensive selection of built-in calculations, including blank subtraction, standard curve creation, classification, quality control, kinetic analysis) and exporting capabilities. Each user can install SkanIt software on his/her own computer to set up assays and analyze data.

Includes:
SkanIt PC Software. Filters available upon request.

Applications include:
Reporter gene assays, immunoassays with luminescent substrates, cell proliferation and cytotoxicity assays, intracellular Ca2+ assays, ATP assays, phagocytosis assays, reactive oxygen assays, microbiological assays, enzyme assays, BRET assays, ADMEtox

TRUSTED_SUSTAINABILITY

Specifications

Compatibility Robot
Detector Type Photomultiplier Tube (PMT)
Dimensions (L x W x H) 420 x 420 x 340 mm (16.5 x 16.5 x 13.4 in.)
Incubator Temperature Ambient +3°C to 45°C
Interface PC Software (Thermo Scientific SkanIt Software)
Measurement Speed 18 s for 96-well Plate
Measurement Technology Luminescence
Plate Type 6 to 384-well Plates
Product Line Luminoskan
Reading Options Top and Bottom
Read-out Range Up to approx. 5000 Relative Light Units (RLU)
Shaking Orbital
Software Type SkanIt Software
Wavelength Selection Filters
Power Consumption 200 VA max.
Shipping Condition Room Temperature
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.