Learn More
Thermo Scientific™ Luminoskan™ Microplate Luminometer, one dispenser
Useful for a variety of luminometric research applications, the Thermo Scientific Luminoskan Microplate Luminometer is a compact and robust instrument offering plate format versatility, fast reading speeds, and up to three dispensers.
Brand: Thermo Scientific™ 5300331
Description
This instrument has passed validation at Promega Corporation to be certified as DLReady. In addition, the Luminoskan Microplate Luminometer is validated for Thermo Scientific Pierce luciferase assays. Luminoskan offers simplicity and reliability for increased efficiency of workflows.
• High sensitivity for both top and bottom reading
• Fast reading speed essential for kinetic applications
• Onboard dispensers for precise initiation of kinetic reactions
• Powerful and easy-to-use Thermo Scientific SkanIt software
High sensitivity for both top and bottom reading
The use of fiberless direct illumination optics for both top and bottom reading ensures high sensitivity, a wide dynamic range, and low cross-talk for accurate and precise results in luminometric assays. The Luminoskan luminometer is certified as DLReady for excellent sensitivity.
Fast reading speed essential for kinetic applications
The Luminoskan Microplate Luminometer reads a 96-well plate in just 18 seconds, essential for kinetic applications such as enzyme kinetic studies and phagocytosis assays.
Onboard dispensers for precise initiation of kinetic reactions
The Luminoskan Microplate Luminometer can be equipped with up to three reagent dispensers* for use during fast kinetic assays, such as Ca2+ flux assays. The onboard dispensers support simultaneous dispensing and reading, enabling the monitoring of fast kinetic measurements form the very start of the reaction. Very low dead volume and backflush capability support conservation of reagents.
*Upgrade to third dispenser performed by service technician
Powerful and easy-to-use SkanIt software
With a visual workflow setup, SkanIt software is quick and easy to learn. It offers effortless data analysis (with a comprehensive selection of built-in calculations, including blank subtraction, standard curve creation, classification, quality control, kinetic analysis) and exporting capabilities. SkanIt software is an open license platform that allows unlimited installations.
Includes:
SkanIt PC Software. Filters available upon request.
Applications include:
Reporter gene assays, immunoassays with luminescent substrates, cell proliferation and cytotoxicity assays, intracellular Ca2+ assays, ATP assays, phagocytosis assays, reactive oxygen assays, microbiological assays, enzyme assays, BRET assays, ADMEtox
Specifications
Robot | |
420 x 420 x 340 mm (16.5 x 16.5 x 13.4 in.) | |
PC Software (Thermo Scientific SkanIt Software) | |
Luminescence | |
Luminoskan™ | |
Up to approx. 5000 Relative Light Units (RLU) | |
SkanIt Software | |
200 VA max. |
Photomultiplier Tube (PMT) | |
Ambient +3°C to 45°C | |
18 s for 96-well Plate | |
6- to 384-well Plates | |
Top and Bottom | |
Orbital | |
Filters | |
Room Temperature |