Learn More
Agilent BioTek Luminescence Test Plate
Product Code. 13467143 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
All Cytation and Synergy (Except Synergy HT) Readers
Unit Size:
Each
Product Code. 13467143
Brand: Agilent BioTek 8030015
Luminescence Test Plate is a NIST-traceable luminescence test plate, for use with all Cytation and Synergy (except Synergy HT) readers
  • Requires updated Users Manual or corresponding Product Qualification Package (sold separately) to operate
  • Helps automate key testing as part of your instrument validation program
  • NIST-traceability certificate included, guarantees a controlled light output from the test plate
  • Simple design, easy to use: just turn the plate on, and read the ultra-stable, low light level LEDs
  • Includes spare battery
TRUSTED_SUSTAINABILITY
Specifications
Product Type Luminescence Test Plate
For Use With (Equipment) All Cytation and Synergy (Except Synergy HT) Readers
For Use With (Application) Automate Key Testing in Instrument Validation Programs
Includes NIST-traceability Certificate