Learn More
Cytiva Luer Connectors for Chromatography Columns
For use with the HiTrap column family of columns
Brand:  Cytiva 18-1027-12
Code : GE
Additional Details : Weight : 0.00300kg
 View more versions of this product
Product Code. 11832180
  • £43.14 / Pack of 2
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Requires : Cytiva HiTrap Columns, sold separately.
Description
Description
  • HiTrap column inlets have 1/16 in. female threads, while the outlets have 1/16 in. male threads
Specifications
Specifications
Union
Polypropylene
Connect a syringe to the to p of a HiTrap column
2 Pack
Column Accesory