missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Luciferase Antibody (Luci17), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Luciferase |
|---|---|
| Clone | Luci17 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066974
|
Novus Biologicals™
NBP3-30808-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066927
|
Novus Biologicals™
NBP3-30808-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Luciferase Monoclonal antibody specifically detects Luciferase in Firefly samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| Luciferase | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Firefly | |
| ec 1.13.12.7, firefly luciferase, Fluc, LuC, luciferin 4 monooxygenase, Luciferin 4-monooxygenase | |
| IgG1 | |
| Protein A or G purified |
| Luci17 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with Luciferase protein isolated from Photinus pyralis. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title