missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Luciferase Antibody (Luci17), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30808-100ug
This item is not returnable.
View return policy
Description
Luciferase Monoclonal antibody specifically detects Luciferase in Firefly samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Luciferase | |
| Monoclonal | |
| Unconjugated | |
| ec 1.13.12.7, firefly luciferase, Fluc, LuC, luciferin 4 monooxygenase, Luciferin 4-monooxygenase | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with Luciferase protein isolated from Photinus pyralis. | |
| 100 μg | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| Luci17 | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Firefly | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction