missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTB Monoclonal Antibody (K1E047_3G1), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | LTB |
|---|---|
| Clone | K1E047_3G1 |
| Concentration | 0.66 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30204795
|
Invitrogen™
MA555821 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30195456
|
Invitrogen™
MA556201 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: QGGLVTETAD PGAQAQQGLG FQKLPEEEPE TDLSPGLPAA HLIGAPLKGQ GLGWETTKEQ AFLTSGTQFS DAEGLALPQD GLYYLYCLVG YRGRAPPGGG DPQGRSVTLR SSLYRAGGAY GPGTPELLLE GAETVTPVLD PARRQGYGPL WYTSVGFGGL VQLRRGERVY VNISHPDMVD FARGKTFFGA
LTB Monoclonal antibody specifically detects LTB in Human samples. It is validated for Western BlotSpecifications
| LTB | |
| 0.66 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide | |
| AI662801; LTB; LTbeta; LT-beta; lymphotoxin B; lymphotoxin beta; lymphotoxin beta (TNF superfamily, member 3); lymphotoxin-beta; p33; TNF superfamily member 3; Tnfc; TNF-C; Tnfsf3; TNLG1C; tumor necrosis factor C; tumor necrosis factor ligand 1C; tumor necrosis factor ligand superfamily member 3 | |
| Recombinant human LTB (immunogen range is 51-240). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LTB |
| K1E047_3G1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q06643 | |
| 4050 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title