missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTB Monoclonal Antibody (K1E047_3G1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556201
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: QGGLVTETAD PGAQAQQGLG FQKLPEEEPE TDLSPGLPAA HLIGAPLKGQ GLGWETTKEQ AFLTSGTQFS DAEGLALPQD GLYYLYCLVG YRGRAPPGGG DPQGRSVTLR SSLYRAGGAY GPGTPELLLE GAETVTPVLD PARRQGYGPL WYTSVGFGGL VQLRRGERVY VNISHPDMVD FARGKTFFGA
Lymphotoxin beta is a type II membrane protein of the TNF family. It anchors lymphotoxin-alpha to the cell surface through heterotrimer formation. The predominant form on the lymphocyte surface is the lymphotoxin-alpha 1/beta 2 complex (e.g. 1 molecule alpha/2 molecules beta) and this complex is the primary ligand for the lymphotoxin-beta receptor. The minor complex is lymphotoxin-alpha 2/beta 1. LTB is an inducer of the inflammatory response system and involved in normal development of lymphoid tissue. Lymphotoxin-beta isoform b is unable to complex with lymphotoxin-alpha suggesting a function for lymphotoxin-beta which is independent of lympyhotoxin-alpha. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008].
Specifications
| LTB | |
| Monoclonal | |
| 0.66 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide | |
| Q06643 | |
| LTB | |
| Recombinant human LTB (immunogen range is 51-240). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K1E047_3G1 | |
| Unconjugated | |
| LTB | |
| AI662801; LTB; LTbeta; LT-beta; lymphotoxin B; lymphotoxin beta; lymphotoxin beta (TNF superfamily, member 3); lymphotoxin-beta; p33; TNF superfamily member 3; Tnfc; TNF-C; Tnfsf3; TNLG1C; tumor necrosis factor C; tumor necrosis factor ligand 1C; tumor necrosis factor ligand superfamily member 3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction