missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57082
This item is not returnable.
View return policy
Description
LSM8 Polyclonal specifically detects LSM8 in Human samples. It is validated for Western Blot.
Specifications
| LSM8 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ33242, LSM8 homolog, U6 small nuclear RNA associated (S. cerevisiae), MAK31-like protein, N(alpha)-acetyltransferase 38, NatC auxiliary subunit, N-alpha-acetyltransferase 38, NatC auxiliary subunit, U6 small nuclear RNA associated, YJR022W | |
| Rabbit | |
| 10 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Zebrafish: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O95777 | |
| NAA38 | |
| Synthetic peptides corresponding to LSM8 (N(alpha)-acetyltransferase 38, NatC auxiliary subunit) The peptide sequence was selected from the N terminal of LSM8. Peptide sequence MTSALENYINRTVAVITSDGRMIVGTLKGFDQTINLILDESHERVFSSSQ. | |
| Affinity purified | |
| RUO | |
| 51691 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction