missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM12 Mouse anti-Human, Unconjugated, Clone: 2E3G3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | LSM12 |
|---|---|
| Clone | 2E3G3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16809415
|
Proteintech
67029-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16899405
|
Proteintech
67029-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The LSM12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LSM12. This antibody recognizes human, mouse, rat antigen. The LSM12 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| LSM12 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 124801 | |
| Primary |
| 2E3G3 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title