Learn More
LSM12 Mouse anti-Human, Unconjugated, Clone: 2E3G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67029-1-IG-150UL
Description
The LSM12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LSM12. This antibody recognizes human, mouse, rat antigen. The LSM12 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| LSM12 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry | |
| 2E3G3 | |
| Unconjugated | |
| 150 μL | |
| 124801 | |
| IgG2a |