missing translation for 'onlineSavingsMsg'
Learn More

LRRN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£381.00

Specifications

Antigen LRRN2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18269422
View Documents
Novus Biologicals
NBP1-59796
100 μL
£381.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LRRN2 Polyclonal specifically detects LRRN2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

LRRN2
Polyclonal
Rabbit
O75325
10446
Synthetic peptides corresponding to LRRN2(leucine rich repeat neuronal 2) The peptide sequence was selected from the middle region of LRRN2. Peptide sequence RVPRRALEQVPGLKFLDLNKNPLQRVGPGDFANMLHLKELGLNNMEELVS.
Primary
Western Blot
Unconjugated
RUO
FIGLER7, GAC1LRRN5, Glioma amplified on chromosome 1 protein, immunoglobulin and leucine rich repeat domain 7, leucine rich and ankyrin repeats 1, leucine rich repeat neuronal 2, leucine rich repeat neuronal 5, leucine-rich repeat neuronal protein 2, Leucine-rich repeat neuronal protein 5
LRRN2
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.