missing translation for 'onlineSavingsMsg'
Learn More

LRRN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-59796

 View more versions of this product

Product Code. 18269422

  • £381.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LRRN2 Polyclonal specifically detects LRRN2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRRN2
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
FIGLER7, GAC1LRRN5, Glioma amplified on chromosome 1 protein, immunoglobulin and leucine rich repeat domain 7, leucine rich and ankyrin repeats 1, leucine rich repeat neuronal 2, leucine rich repeat neuronal 5, leucine-rich repeat neuronal protein 2, Leucine-rich repeat neuronal protein 5
Rabbit
Affinity purified
RUO
10446
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
O75325
LRRN2
Synthetic peptides corresponding to LRRN2(leucine rich repeat neuronal 2) The peptide sequence was selected from the middle region of LRRN2. Peptide sequence RVPRRALEQVPGLKFLDLNKNPLQRVGPGDFANMLHLKELGLNNMEELVS.
100 μL
Primary
Expected identity based on immunogen sequence: Bovine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
Human, Mouse, Rat, Pig, Bovine, Canine, Equine
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.