missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRK2 Antibody (S138-6), Novus Biologicals™
Mouse Monoclonal Antibody
£181.00 - £453.00
Specifications
| Antigen | LRRK2 |
|---|---|
| Clone | S138-6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence |
| Applications | Western Blot, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616656
|
Novus Biologicals
NBP2-42175 |
0.1 mg |
£453.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660489
|
Novus Biologicals
NBP2-42175-0.025mg |
0.025 mg |
£181.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRRK2 Monoclonal antibody specifically detects LRRK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| LRRK2 | |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase | |
| PBS (pH 7.4), 50% Glycerol | |
| augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
| Fusion protein, amino acids 1-500 (N-terminus) of human LRRK2. 83% identical in mouse and rat. No significant identity with LRRK1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S138-6 | |
| Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q5S007 | |
| 120892 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title