missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRK2 Antibody (S138-6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42175
This item is not returnable.
View return policy
Description
LRRK2 Monoclonal antibody specifically detects LRRK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| LRRK2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence | |
| Q5S007 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| S138-6 | |
| Unconjugated | |
| PBS (pH 7.4), 50% Glycerol | |
| augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
| Fusion protein, amino acids 1-500 (N-terminus) of human LRRK2. 83% identical in mouse and rat. No significant identity with LRRK1. | |
| 0.1 mg | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase | |
| 120892 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction