missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRK2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 39 publications
£141.00 - £417.00
Specifications
| Antigen | LRRK2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:2000-1:5000, Flow Cytometry 1:200, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1000-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen, Knockout Validated |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18044085
|
Novus Biologicals
NB300-268SS |
0.025 mL |
£141.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18717563
|
Novus Biologicals
NB300-268 |
0.1 mL |
£417.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRRK2 Polyclonal specifically detects LRRK2 in Human, Mouse, Bovine, C. elegans, Insect, Plant samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated.Specifications
| LRRK2 | |
| Western Blot 1:2000-1:5000, Flow Cytometry 1:200, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1000-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen, Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| PBS with 0.1% Sodium Azide | |
| augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
| LRRK2 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, bovine, S. cerevisiae, Drosophila and moth. Immunogen sequence has 87% homology to rat. |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Bovine, C. elegans, Insect, Plant | |
| Q5S007 | |
| 120892 | |
| A C-terminal synthetic peptide made to the human LRRK2 protein sequence (between residues 2500-2527). [UniProt# Q5S007] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 286 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title