missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRK2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 39 publications
Brand: Novus Biologicals NB300-268
This item is not returnable.
View return policy
Description
LRRK2 Polyclonal specifically detects LRRK2 in Human, Mouse, Bovine, C. elegans, Insect, Plant samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated.
Specifications
| LRRK2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.1% Sodium Azide | |
| augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
| Rabbit | |
| 286 kDa | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 120892 | |
| Human, Mouse, Bovine, C. elegans, Insect, Plant | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
| 1 mg/mL | |
| Western Blot 1:2000-1:5000, Flow Cytometry 1:200, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:1000-1:2000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen, Knockout Validated | |
| Q5S007 | |
| LRRK2 | |
| A C-terminal synthetic peptide made to the human LRRK2 protein sequence (between residues 2500-2527). [UniProt# Q5S007] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, bovine, S. cerevisiae, Drosophila and moth. Immunogen sequence has 87% homology to rat. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction