missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC73 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56940
This item is not returnable.
View return policy
Description
LRRC73 Polyclonal specifically detects LRRC73 in Human samples. It is validated for Western Blot.
Specifications
| C6orf154 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C6orf154, chromosome 6 open reading frame 154, dJ337H4.2, FLJ44836, hypothetical protein LOC221424, leucine rich repeat containing 73, MGC131686 | |
| Rabbit | |
| 33 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q5JTD7 | |
| LRRC73 | |
| Synthetic peptide directed towards the N terminal of human C6orf154 (NP_001012992). Peptide sequence MLPSSIQISGEPLSGAEVRDICRGLRDNAVRLLSLRGCRLCDRDFGRICR. | |
| Affinity purified | |
| RUO | |
| 221424 | |
| Human, Mouse, Rat, Bovine, Guinea Pig, Goat, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction