missing translation for 'onlineSavingsMsg'
Learn More

LRAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen LRAT
Concentration 0.27 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16890724
View Documents
Proteintech
12815-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16880724
View Documents
Proteintech
12815-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody was generated against the internal residues of human LRAT. It is predicted to detect endogenous LRAT.

Lecithin retinol acyltransferase (LRAT) is a membrane bound enzyme that catalyzes the transfer of an acyl group from the sn-1 position of lecithin to vitamin A which generates all-trans-retinyl esters (tREs) in the liver, some extrahepatic tissues, such as the lung, and retinal pigmented epithelium. LRAT can also exchange palmitoyl groups between RPE65, a tRE binding protein essential for vision, and tREs, which is important for the operation of the visual pathway. LRAT is essential for the dietary mobilization, transport, and storage of vitamin A as well as the synthesis of the visual pigment chromophore. LRAT monomers interact in membranes to form homodimers through disulfide bond formation. A loss of LRAT correlates with an early onset severe retinal dystrophy and severe retinyl ester deprivation, while a reduction in LRAT expression may be associated with invasive bladder cancer.
Specifications

Specifications

LRAT
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
O95237, Q9JI60, Q9JI61
64047, 79235, 9227
LRAT Fusion Protein Ag3556
Primary
-20°C
LRAT
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
LRAT
Lrat
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.