Learn More
LRAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12815-1-AP-20UL
 View more versions of this product
Product Code. 16890724
  • £135.00 / 20µL
Description
This antibody was generated against the internal residues of human LRAT. It is predicted to detect endogenous LRAT.
Lecithin retinol acyltransferase (LRAT) is a membrane bound enzyme that catalyzes the transfer of an acyl group from the sn-1 position of lecithin to vitamin A which generates all-trans-retinyl esters (tREs) in the liver, some extrahepatic tissues, such as the lung, and retinal pigmented epithelium. LRAT can also exchange palmitoyl groups between RPE65, a tRE binding protein essential for vision, and tREs, which is important for the operation of the visual pathway. LRAT is essential for the dietary mobilization, transport, and storage of vitamin A as well as the synthesis of the visual pigment chromophore. LRAT monomers interact in membranes to form homodimers through disulfide bond formation. A loss of LRAT correlates with an early onset severe retinal dystrophy and severe retinyl ester deprivation, while a reduction in LRAT expression may be associated with invasive bladder cancer.
Specifications
Specifications
LRAT
Polyclonal
Unconjugated
LRAT
LRAT
Rabbit
Antigen Affinity Chromatography
RUO
64047, 79235, 9227
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95237, Q9JI60, Q9JI61
Lrat
LRAT Fusion Protein Ag3556
20 μL
Primary
Human, Mouse, Rat