missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPAR6/P2RY5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | LPAR6/P2RY5 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
LPAR6/P2RY5 Polyclonal specifically detects LPAR6/P2RY5 in Human samples. It is validated for Western Blot.Specifications
| LPAR6/P2RY5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS buffer, 2% sucrose | |
| 10161 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| G-protein coupled purinergic receptor P2Y5, LAH3, LPA receptor 6, LPA-6, lysophosphatidic acid receptor 6, MGC120358, Oleoyl-L-alpha-lysophosphatidic acid receptor, P2RY5ARWH1, P2Y purinoceptor 5, P2Y5RB intron encoded G-protein coupled receptor, Purinergic receptor 5, purinergic receptor P2Y, G-protein coupled, 5 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human LPAR6/P2RY5 (NP_005758). Peptide sequence GDLLCKISVMLFYTNMYGSILFLTCISVDRFLAIVYPFKSKTLRTKRNAK | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title