missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LPAR3/LPA3/EDG-7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£208.00 - £290.00
Specifications
| Antigen | LPAR3/LPA3/EDG-7 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:40000, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041734
|
Novus Biologicals™
NBP3-27861-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041607
|
Novus Biologicals™
NBP3-27861-50ul |
50 μL |
£208.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LPAR3/LPA3/EDG-7 Polyclonal antibody specifically detects LPAR3/LPA3/EDG-7 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| LPAR3/LPA3/EDG-7 | |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS, 50% glycerol, 0.5% BSA | |
| 23566 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot 1:500 - 1:2000, ELISA 1:40000, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Edg-7, EDG7FLJ98231, GPCR, HOFNH30, LPA-3, LPA3LPA receptor 3, lysophosphatidic acid G-protein-coupled receptor7,LPA receptor EDG7, lysophosphatidic acid receptor 3, Lysophosphatidic acid receptor Edg-7 | |
| The antiserum was produced against synthesized peptide derived from human LPAR3/LPA3/EDG-7 (Uniprot #: Q9UBY5) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title