missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LPAR3/LPA3/EDG-7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-27861-50ul

 View more versions of this product

Product Code. 30041607

  • £208.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LPAR3/LPA3/EDG-7 Polyclonal antibody specifically detects LPAR3/LPA3/EDG-7 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LPAR3/LPA3/EDG-7
Polyclonal
Western Blot 1:500 - 1:2000, ELISA 1:40000, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000
Edg-7, EDG7FLJ98231, GPCR, HOFNH30, LPA-3, LPA3LPA receptor 3, lysophosphatidic acid G-protein-coupled receptor7,LPA receptor EDG7, lysophosphatidic acid receptor 3, Lysophosphatidic acid receptor Edg-7
The antiserum was produced against synthesized peptide derived from human LPAR3/LPA3/EDG-7 (Uniprot #: Q9UBY5)
50 μL
GPCR
23566
Store at -20°C. Avoid freeze/thaw cycles.
IgG
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS, 50% glycerol, 0.5% BSA
Rabbit
Antigen Affinity-purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.