missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LPAR3/LPA3/EDG-7 Antibody - C-terminus, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £591.00
Specifications
| Antigen | LPAR3/LPA3/EDG-7 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067215
|
Novus Biologicals™
NBP3-30810-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066928
|
Novus Biologicals™
NBP3-30810-100ug |
100 μg |
£591.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LPAR3/LPA3/EDG-7 Polyclonal antibody specifically detects LPAR3/LPA3/EDG-7 in Human samples. It is validated for Western BlotSpecifications
| LPAR3/LPA3/EDG-7 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Edg-7, EDG7FLJ98231, GPCR, HOFNH30, LPA-3, LPA3LPA receptor 3, lysophosphatidic acid G-protein-coupled receptor7,LPA receptor EDG7, lysophosphatidic acid receptor 3, Lysophosphatidic acid receptor Edg-7 | |
| Antibody raised against a unique synthetic peptide derived from the C-terminal of the human LPAR3/LPA3/EDG-7 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| Phosphate buffered saline | |
| 23566 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title