missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LPAR3/LPA3/EDG-7 Antibody - C-terminus, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30810-25ug
This item is not returnable.
View return policy
Description
LPAR3/LPA3/EDG-7 Polyclonal antibody specifically detects LPAR3/LPA3/EDG-7 in Human samples. It is validated for Western Blot
Specifications
| LPAR3/LPA3/EDG-7 | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Edg-7, EDG7FLJ98231, GPCR, HOFNH30, LPA-3, LPA3LPA receptor 3, lysophosphatidic acid G-protein-coupled receptor7,LPA receptor EDG7, lysophosphatidic acid receptor 3, Lysophosphatidic acid receptor Edg-7 | |
| Antibody raised against a unique synthetic peptide derived from the C-terminal of the human LPAR3/LPA3/EDG-7 protein. | |
| 25 μg | |
| GPCR | |
| 23566 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction