missing translation for 'onlineSavingsMsg'
Learn More

Loxoprofen L-Methol Ester, TRC

Product Code. 30467676
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467676 250 mg 250mg
30467229 100 mg 100mg
30479825 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30467676 Supplier Toronto Research Chemicals Supplier No. TRCM305190250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1091621-63-4
Molecular Formula C25H36O3
Molecular Weight (g/mol) 384.56
Synonym Loxoprofen Related Compound 14, alpha-Methyl-4-[(2-oxocyclopentyl)methyl]-benzeneacetic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester
IUPAC Name [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2-[4-[(2-oxocyclopentyl)methyl]phenyl]propanoate
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)C(C)c2ccc(CC3CCCC3=O)cc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C25H36O3/c1-16(2)22-13-8-17(3)14-24(22)28-25(27)18(4)20-11-9-19(10-12-20)15-21-6-5-7-23(21)26/h9-12,16-18,21-22,24H,5-8,13-15H2,1-4H3/t17-,18?,21?,22+,24-/m1/s1
Formula Weight 384.2664
Chemical Name or Material Loxoprofen L-Methol Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.