CAS RN 1091621-63-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Loxoprofen L-Methol Ester, TRC
CAS: 1091621-63-4 Molecular Formula: C25H36O3 Molecular Weight (g/mol): 384.56 Synonym: Loxoprofen Related Compound 14,alpha-Methyl-4-[(2-oxocyclopentyl)methyl]-benzeneacetic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester IUPAC Name: [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2-[4-[(2-oxocyclopentyl)methyl]phenyl]propanoate SMILES: CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)C(C)c2ccc(CC3CCCC3=O)cc2