missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00
Specifications
| Antigen | LOX |
|---|---|
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 29404848), Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen, Knockout Validated reported in scientific literature (PMID 31173582) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
LOX Polyclonal specifically detects LOX in Human, Mouse, Rat, Porcine, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifications
| LOX | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
| 0.1 mL crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| 4015 | |
| A portion of amino acids 305-338 of human LOX protein were used as the immunogen. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 29404848), Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen, Knockout Validated reported in scientific literature (PMID 31173582) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Pig, Sheep | |
| lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
| LOX | |
| IgG | |
| Unpurified | |
| This antibody is reactive to Bovine, Chimpanzee, Human, Mouse and Rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title