missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24877SS
This item is not returnable.
View return policy
Description
LOX Polyclonal specifically detects LOX in Human, Mouse, Rat, Porcine, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.
Specifications
| LOX | |
| Polyclonal | |
| Western Blot 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 29404848), Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen, Knockout Validated reported in scientific literature (PMID 31173582) | |
| lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| This antibody is reactive to Bovine, Chimpanzee, Human, Mouse and Rat. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.1 mL crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| LOX | |
| A portion of amino acids 305-338 of human LOX protein were used as the immunogen. | |
| 0.025 mL | |
| Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
| 4015 | |
| Human, Mouse, Rat, Pig, Sheep | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction