missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Lower Part For Research 3 Neo Single Channel Pipettes

Product Code. 30689640 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
2.5 μL
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
2 mL
5 mL
10 mL
Color:
Dark Gray
Light Gray
Turquoise
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color unitSize
30689640 2.5 μL Dark Gray Each
30689733 10 μL - Each
30689633 20 μL Light Gray Each
30689671 20 μL - Each
30689727 100 μL - Each
30689723 200 μL - Each
30689731 300 μL - Each
30689685 1000 μL - Each
30689657 2 mL - Each
30689667 5 mL - Each
30689691 10 mL Turquoise Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 30689640 Supplier Eppendorf™ Supplier No. 3102634000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This original Eppendorf spare part is designed to replace the entire lower part including the piston cone

  • Eppendorf spare part is designed to replace the entire lower part including the piston cone, of Eppendorf Research™
  • 3 neo pipettes, which is housed inside the ejector sleeve
  • Available in various color code and Volume capacity
TRUSTED_SUSTAINABILITY

Specifications

Color Dark Gray
For Use With Pipettes
Material Thermoplastic
Type Pipette Lower Part
Volume (Metric) 2.5 μL
For Use With (Equipment) Pipette Tips
Quantity 1 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.