missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Eppendorf spare part is designed to replace the entire lower part including the piston cone, of Eppendorf Research™
- 3 neo pipettes, which is housed inside the ejector sleeve
- Available in various color code and Volume capacity
Specifications
Specifications
| Color | Light Gray |
| For Use With | Pipettes |
| Material | Thermoplastic |
| Type | Pipette Lower Part |
| Volume (Metric) | 20 μL |
| For Use With (Equipment) | Pipette Tips |
| Quantity | 1 Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?