missing translation for 'onlineSavingsMsg'
Learn More

Lovastatin, 97%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar H52792.06

Quantity 5g

 View more versions of this product

Product Code. 15489537

  • £79.10 / 5g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Lovastatin is a cholesterol lowering drug and competitive inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. It increases cellular resistance to anticancer agents such as doxorubicin and induces apoptosis in myeloma cells. It causes cell cycle arrest in G1 and G2/M phases.

Solubility
Soluble in DMSO (10 mg/ml), ethanol (10 mg/ml), methanol, chloroform, acetone, acetonitrile, DMF, and water (0.4 µg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

75330-75-5
404.55
PCZOHLXUXFIOCF-BXMDZJJMSA-N
53232
(1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate
C24H36O5
MFCD00072164
lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin
CHEBI:40303
CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

75330-75-5
MFCD00072164
lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin
PCZOHLXUXFIOCF-BXMDZJJMSA-N
(1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate
53232
404.56
Lovastatin
C24H36O5
5g
Soluble in DMSO (10mg/ml),ethanol (10mg/ml),methanol,chloroform,acetone,acetonitrile,DMF,and water (0.4 μg/ml).
CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12
404.55
CHEBI:40303
97%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.