Learn More
Fisherbrand™ Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer
Product Code. 15577603 Shop All Fisherbrand Products
Click to view available options
Data Memory:
7680 Readings
Unit Size:
Each
Product Code. 15577603
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Requires
Requires MaxiThermal software (15-081-150) and cradle with USB cable (14-650-123) to download data and restart mini datalogger.
Know the temperature excursion duration immediately, no data download required with Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer.
  • Duration of Temperature excursion
  • Data-logger with user-defined recording intervals
  • Active LCD Display shows the following important data: Current temperature, Minimum/Maximum temperatures, Recording status, Run Time, Alarm (Excursion) Cumulative Time, Alarm Tally
  • Requires the Fisherbrand Traceable Cradle and USB Cable (6549) to upload data to PC
USER SELECTABLE RECORDING INTERVALS For complete flexibility, the user-defined recording interval may be set based on the number of desired days to log (from 4 hours to 365 days) or the desired time interval (from every 2 seconds to every 24 hours).Internal flash memory stores 7680 readings.
To ensure that no data is lost, once the memory is full the thermometer will stop logging data (data will not be overwritten).While recording, the following information is displayed: Current, Minimum and Maximum temperature achieved and cumulative running time. Once an alarm condition has occurred, the following additional information is displayed: Alarm Tally, ALARM notification, cumulative alarm time.
Individual Logger-Trac Thermometers are pre-programmed with alarm values of 2 °C (low) and 8 °C (high) and a recording interval to allow for 30 days of recording (readings are logged every 5 minutes, 33 seconds).
Specifications
Type Datalogger
Display Type LCD
Data Memory 7680 Readings
Calibration Not Calibrated
Alarm Types High/Low
Software Type TraceableLIVE
Description With temperature MIN/MAX memory. data logging and exportable data
Interface USB
IP Rating IP66
Sensor Types Internal Sensor
For Use With (Application) Perfect solution for monitoring material in transit as well as reagents, clinical samples, vaccines, pharmaceuticals and food items in any location
Accuracy (Temperature) ±0.2°C (0°C; to 10°C;), ±1°C otherwise
Temperature Range (English) -31°F to +122°F
Temperature Range (Metric) -35°C to +50°C
Dimensions (L x W x H) 2 x 1 x 0.5 in.
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt