Learn More
Eppendorf™ Locking Clip
Prevent spring action of tip cones with these locking clips
Brand: Eppendorf™ 3122612006
Code : NEW
Additional Details : Weight : 0.01100kg
Description
- Accessory for Eppendorf Xplorer™/Xplorer plus
- Locking clip
- Available for 8- and 12-channel pipettes
Specifications
Locking clip | |
Preventing spring action of tip cones | |
8-channel pipette | |
1 Each |