Learn More
Eppendorf™ Locking Clip
Prevent spring action of tip cones with these locking clips
£6.30 - £7.70
Specifications
Type | Locking clip |
---|---|
For Use With (Application) | Preventing spring action of tip cones |
Quantity | 1 Each |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15204326
|
Eppendorf™
3122612006 |
8-channel pipette |
£6.30
Each |
In Stock
Log in to see stock. |
|||||
15234326
|
Eppendorf™
3122613002 |
12-channel pipette |
£7.70
Each |
In Stock
Log in to see stock. |
|||||
Description
- Accessory for Eppendorf Xplorer™/Xplorer plus
- Locking clip
- Available for 8- and 12-channel pipettes
Specifications
Locking clip | |
Preventing spring action of tip cones | |
1 Each |