missing translation for 'onlineSavingsMsg'
Learn More

Lobetyolin, TRC

Product Code. 30479875
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479875 1 mg 1mg
30467905 5 mg 5mg
30473234 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479875 Supplier Toronto Research Chemicals Supplier No. TRCL4693551MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136085-37-5
Molecular Formula C20H28O8
Molecular Weight (g/mol) 396.43
Synonym (7E)-2-Hydroxy-1-[(1E)-5-hydroxy-1-pentenyl]-7-nonene-3,5-diynyl β-D-glucopyranoside, (7E)-2-Hydroxy-1-[(1E)-5-hydroxy-1-penten-1-yl]-7-nonene-3,5-diyn-1-yl β-D-Glucopyranoside,
SMILES OCC1[C@H](O)C(O)C(O)[C@@H](O1)OC2[C@@H](O)C(CO)O[C@@H](O)C2O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H28O8/c1-2-3-4-5-7-10-14(23)15(11-8-6-9-12-21)27-20-19(26)18(25)17(24)16(13-22)28-20/h2-3,8,11,14-26H,6,9,12-13H2,1H3/b3-2+,11-8+/t14?,15?,16-,17-,18+,19-,20-/m1/s1
Chemical Name or Material Lobetyolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.